Learn More
Aviva Systems Biology Corp Zfr Antibody - N-terminal region (ARP39226_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39226_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zfr. It was validated on Western Blot. . Alternate names: C920030H05Rik, MGC176100ApplicationWB, IHC. Species reactivity: Dog, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-789