Learn More
Cayman Chemical Z-FR-AMC (trifluoroacetate salt), Purity: ≥98%, 50 mg
SDP
Catalog No. 502950415
Encompass_Preferred
A fluorogenic substrate for cathepsin L; AMC is released upon enzymatic cleavage by cathepsin L and its fluorescence can be used to quantify cathepsin L activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-295-0415 Supplier Cayman Chemical Supplier No. 3404550 MG
A fluorogenic substrate for cathepsin L; AMC is released upon enzymatic cleavage by cathepsin L and its fluorescence can be used to quantify cathepsin L activity; ex/em = 340-360/440-460 nm, respectively