Learn More
Aviva Systems Biology Corp ZFX Antibody - N-terminal region (ARP38309_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38309_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFX. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZFX. Alternate names: ZNF926ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-729