Learn More
Aviva Systems Biology Corp ZFX Antibody - middle region (ARP33498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZFX. Alternate names: ZNF926ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55045