Learn More
Aviva Systems Biology Corp ZFY1 Antibody - middle region (ARP37003_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37003_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFY1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse ZFY1. Alternate names: Zfy-1ApplicationWB. Species reactivity: Mouse
Catalog No. 50-014-56310