Learn More
Aviva Systems Biology Corp ZFYVE1 Antibody - C-terminal region (ARP50843_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50843_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFYVE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZFYVE1. Alternate names: DFCP1, KIAA1589, TAFF1, ZNFN2A1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60448