Learn More
Aviva Systems Biology Corp ZFYVE19 Antibody - C-terminal region (ARP50452_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50452_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFYVE19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZFYVE19. Alternate names: FLJ14840, MPFYVEApplicationWB. Species reactivity: Dog, Guinea Pig, Human
Catalog No. 50-014-60348