Learn More
Aviva Systems Biology Corp RBSN Antibody - N-terminal region (ARP39547_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39547_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFYVE20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZFYVE20. Alternate names: FLJ34993, MGC126210, Rabenosyn-5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57080