Learn More
Aviva Systems Biology Corp ZFYVE27 Antibody - C-terminal region (ARP44429_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44429_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFYVE27. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZFYVE27. Alternate names: RP11-459F3.2, SPG33, PROTRUDINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58806