Learn More
Aviva Systems Biology Corp ZG16 Antibody - middle region (ARP52901_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52901_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZG16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZG16. Alternate names: MGC34820, JCLN, JCLN1, ZG16A, hZG16ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-60864