Learn More
Medchemexpress LLC Zymogen granule protein 16 homolog B | >95.0% | 250UG
SDP
Catalog No. 5000176472
Encompass_Preferred
Recombinant human ZG16B protein (mature chain G17-R172) produced in HEK293 cells and supplied as a lyophilized, C-terminal His-tagged protein for research use. Intended for biochemical characterization, antibody and assay development; reconstitute according to instructions and store frozen in aliquots for long-term stability.
  • Expressed in HEK293 cells (human)
  • Includes C-terminal His tag for purification
  • High purity (>95% by reducing SDS-PAGE)
  • Lyophilized formulation with trehalose for enhanced stability
  • Available in research sizes including 250 μg
  • Store lyophilized at -20 °C; aliquot and freeze at -20 °C or -80 °C after reconstitution
Catalog No. 50-001-76472 Supplier Medchemexpress LLC Supplier No. HYP700451250UG
Recombinant human ZG16B protein (mature chain G17-R172) produced in HEK293 cells and supplied as a lyophilized, C-terminal His-tagged protein for research use. Intended for biochemical characterization, antibody and assay development; reconstitute according to instructions and store frozen in aliquots for long-term stability.
  • Expressed in HEK293 cells (human)
  • Includes C-terminal His tag for purification
  • High purity (>95% by reducing SDS-PAGE)
  • Lyophilized formulation with trehalose for enhanced stability
  • Available in research sizes including 250 μg
  • Store lyophilized at -20 °C; aliquot and freeze at -20 °C or -80 °C after reconstitution