Learn More
Aviva Systems Biology Corp ZGPAT Antibody - middle region (ARP42725_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42725_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZGPAT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZGPAT. Alternate names: GPATC6, GPATCH6, KIAA1847, MGC44880, ZC3H9, ZC3HDC9, ZIP, RP4-583P15.3ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58318