Learn More
Beckman Coulter Ziath DP5 Express Rack Reader - Camera-based 2D DataMatrix rack/tube scanner, USB or Ethernet, compatible with 24-384 position SBS racks
Catalog No. NC3865453
Encompass_Preferred
The Ziath DP5 Express Rack Reader is a compact, camera-based 2D DataMatrix scanner engineered for rapid and reliable reading of barcoded tubes in SBS-format racks. It offers fast imaging, easy integration, and an optional cryoprotection coating for cryogenic use.
  • Fast image capture with approximately one second per scan.
  • Reads 24, 48, 96, 240, and 384 position SBS-format racks.
  • Connects via USB or Ethernet (RJ45) for flexible integration.
  • Small footprint suitable for robotic workflows and limited benchtop space.
  • Optional passive cryoprotection coating prevents condensation during cryogenic scanning.
  • Includes software with remote control and multiple export formats.
  • Compatible with Windows 10 and 11 and backed by a two-year warranty.
Catalog No. NC3865453 Supplier Beckman Coulter Supplier No. D19329
Please call customer service for current pricing. 1-800-766-7000
The Ziath DP5 Express Rack Reader is a compact, camera-based 2D DataMatrix scanner engineered for rapid and reliable reading of barcoded tubes in SBS-format racks. It offers fast imaging, easy integration, and an optional cryoprotection coating for cryogenic use.
  • Fast image capture with approximately one second per scan.
  • Reads 24, 48, 96, 240, and 384 position SBS-format racks.
  • Connects via USB or Ethernet (RJ45) for flexible integration.
  • Small footprint suitable for robotic workflows and limited benchtop space.
  • Optional passive cryoprotection coating prevents condensation during cryogenic scanning.
  • Includes software with remote control and multiple export formats.
  • Compatible with Windows 10 and 11 and backed by a two-year warranty.