Learn More
Beckman Coulter Ziath DP5 Express Rack Reader - Compact 2D datamatrix tube rack reader for 24-384 well SBS racks
Catalog No. NC3866467
Encompass
The DP5 Express Rack Reader is a compact, camera-based 2D datamatrix reader that captures images in approximately one second and is suitable for integration with automated workflows. It supports multiple SBS-format rack sizes and can be fitted with optional cryoprotection to prevent condensation when scanning cold or cryogenic samples.
  • Camera-based imaging with approximately 1 second capture time.
  • Compatible with 24, 48, 96, 240, and 384 well SBS-format racks.
  • Scans 2D barcoded tubes from common manufacturers.
  • Compact footprint suitable for robotic and automated integration.
  • Optional cryoprotection prevents condensation when scanning cold samples.
Catalog No. NC3866467 Supplier Beckman Coulter Supplier No. D19330
Add to Cart
Add to Cart
Request a Quote
The DP5 Express Rack Reader is a compact, camera-based 2D datamatrix reader that captures images in approximately one second and is suitable for integration with automated workflows. It supports multiple SBS-format rack sizes and can be fitted with optional cryoprotection to prevent condensation when scanning cold or cryogenic samples.
  • Camera-based imaging with approximately 1 second capture time.
  • Compatible with 24, 48, 96, 240, and 384 well SBS-format racks.
  • Scans 2D barcoded tubes from common manufacturers.
  • Compact footprint suitable for robotic and automated integration.
  • Optional cryoprotection prevents condensation when scanning cold samples.