Learn More
Aviva Systems Biology Corp ZIC4 Antibody - N-terminal region (ARP39680_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39680_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZIC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZIC4. Alternate names: FLJ42609, FLJ45833ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-57135