Learn More
Aviva Systems Biology Corp ZIC5 Antibody - N-terminal region (ARP33669_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33669_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZIon Channel5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZIC5. Alternate names: ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Zebrafish
Catalog No. 50-014-55111