Learn More
TARGETMOL CHEMICALS INC Ziconotide Acetate 107452-89-1
SDP
Catalog No. 502880422
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ziconotide Acetate (107452-89-1 free base) (Prialt) is an analgesic agent and has been used to treat neuropathic and non-neuropathic pain. Ziconotide Acetate (107452-89-1 free base) acts by binding to N-type calcium channels situated on the terminal part of primary afferent neurons of the nociceptive pathway therefore reducing synaptic transmission with potent antinociceptive effects. Purity 99.82%
Catalog No. 50-288-0422 Supplier TARGETMOL CHEMICALS INC Supplier No. TP1559L1MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ziconotide Acetate (107452-89-1 free base) (Prialt) is an analgesic agent and has been used to treat neuropathic and non-neuropathic pain. Ziconotide Acetate (107452-89-1 free base) acts by binding to N-type calcium channels situated on the terminal part of primary afferent neurons of the nociceptive pathway therefore reducing synaptic transmission with potent antinociceptive effects. Purity 99.82%