Learn More
Medchemexpress LLC HY-P0062B 5mg , Ziconotide (acetate) CAS:914454-03-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0062B5MG
Encompass_Preferred
HY-P0062B 5mg Ziconotide (acetate) CAS: 914454-03-8 Ziconotide acetate (SNX-111 acetate), a peptide, is a potent and selective block of N-type calcium channels antagonist. Ziconotide acetate reduces synaptic transmission, and can be used for chronic pain research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1979