Learn More
Medchemexpress LLC Ziconotide acetate (SNX-111 acetate) | 914454-03-8 | 99.2% | 2639.13 (free base) | 10 MG
SDP
Catalog No. 5000425747
Encompass_Preferred
Ziconotide acetate is a peptide that acts as a potent and selective antagonist of N-type calcium channels. It reduces synaptic transmission and is suitable for chronic pain research.
  • Potent and selective N-type calcium channel antagonist
  • Reduces synaptic transmission
  • Used for chronic pain research
  • Partially reduces high-voltage-activated calcium currents
  • Delivers antinociceptive efficacy by reducing pronociceptive neurotransmitters
Catalog No. 50-004-25747 Supplier Medchemexpress LLC Supplier No. HYP0062B10MG
Ziconotide acetate is a peptide that acts as a potent and selective antagonist of N-type calcium channels. It reduces synaptic transmission and is suitable for chronic pain research.
  • Potent and selective N-type calcium channel antagonist
  • Reduces synaptic transmission
  • Used for chronic pain research
  • Partially reduces high-voltage-activated calcium currents
  • Delivers antinociceptive efficacy by reducing pronociceptive neurotransmitters