Learn More
Biomatik Corporation Ziconotide (Zic) ELISA Kit
Supplier:  Biomatik Corporation EKU11593
Encompass_Preferred
Ziconotide (Zic) ELISA Kit for detecting Zic in biological fluids. Species Reactivity: Universal. Detection Range: 3.70-300ng/ml. Sensitivity: 1.39ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: SNX-111; Prialt. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7499