Learn More
Medchemexpress LLC Zidebactam | 1436861-97-0 | 100 MG
SDP
Catalog No. 5000233892
Encompass_Preferred
Zidebactam is a beta-lactamase inhibitor and a penicillin-binding protein 2 (PBP2) inhibitor supplied for laboratory research use. It is used to study beta-lactam resistance and to evaluate combination antibiotic therapies.
  • Acts as a beta-lactamase inhibitor and PBP2 inhibitor.
  • Reported PBP2 IC50 ≈ 0.26 μg/mL, suitable for potency benchmarking.
  • Intended for in vitro research applications; not for human use.
  • Useful for studying antibiotic resistance mechanisms and adjuvant therapies.
  • Characterized by CAS 1436861-97-0 and molecular weight 391.40 g/mol.
Catalog No. 50-002-33892 Supplier Medchemexpress LLC Supplier No. HY120859100MG
Zidebactam is a beta-lactamase inhibitor and a penicillin-binding protein 2 (PBP2) inhibitor supplied for laboratory research use. It is used to study beta-lactam resistance and to evaluate combination antibiotic therapies.
  • Acts as a beta-lactamase inhibitor and PBP2 inhibitor.
  • Reported PBP2 IC50 ≈ 0.26 μg/mL, suitable for potency benchmarking.
  • Intended for in vitro research applications; not for human use.
  • Useful for studying antibiotic resistance mechanisms and adjuvant therapies.
  • Characterized by CAS 1436861-97-0 and molecular weight 391.40 g/mol.