Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zidebactam | 1436861-97-0 | 98.6% | 50 MG
SDP

Catalog No. 5000232842
Encompass_Preferred

Zidebactam is a β-lactamase inhibitor and penicillin-binding protein 2 (PBP2) inhibitor used as a research compound and antibiotic adjuvant. CAS 1436861-97-0; molecular weight 391.40 g/mol; supplied as a white-to-yellow solid in a 50 mg vial. Reported activity includes PBP2 inhibition with an IC50 of 0.26 μg/mL. For research use only; not for human or clinical use.

  • Beta-lactamase and PBP2 inhibitor; enhances β-lactam activity.
  • Reported PBP2 IC50 of 0.26 μg/mL against P. aeruginosa PAO1.
  • Molecular weight 391.40 g/mol; formula C13H21N5O7S.
  • Purity 98.6% (reported) and supplied as a solid.
  • Intended for laboratory research use only, not for human use.

Catalog No. 50-002-32842 Supplier Medchemexpress LLC Supplier No. HY12085950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zidebactam is a β-lactamase inhibitor and penicillin-binding protein 2 (PBP2) inhibitor used as a research compound and antibiotic adjuvant. CAS 1436861-97-0; molecular weight 391.40 g/mol; supplied as a white-to-yellow solid in a 50 mg vial. Reported activity includes PBP2 inhibition with an IC50 of 0.26 μg/mL. For research use only; not for human or clinical use.

  • Beta-lactamase and PBP2 inhibitor; enhances β-lactam activity.
  • Reported PBP2 IC50 of 0.26 μg/mL against P. aeruginosa PAO1.
  • Molecular weight 391.40 g/mol; formula C13H21N5O7S.
  • Purity 98.6% (reported) and supplied as a solid.
  • Intended for laboratory research use only, not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.