Learn More
Selleck Chemical LLC Zidovudine 25mg 30516-87-1 Azidothymidine, NSC 602670
SDP
Supplier:  Selleck Chemical LLC S257925MG
Encompass_Preferred
Zidovudine (ZDV, Azidothymidine, NSC 602670) is a nucleoside analogue reverse transcriptase inhibitor, used to treat HIV. It could decrease the HDR efficiency and decrease CRISPR-mediated sequence-specific genome knockin events while increaseing knockout efficiency. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8010