Learn More
Medchemexpress LLC HY-17413 500mg , Zidovudine Azidothymidine;AZT;ZDV CAS:30516-87-1 Purity:98%
SDP
Catalog No. 501492479
Encompass_Preferred
Medchemexpress, HY-17413 500mg Zidovudine Azidothymidine;AZT;ZDV CAS:30516-87-1 Formula:C10H13N5O4 NRTI, HIV CRISPR/Cas9 Purity:98% Zidovudine is a nucleoside reverse transcriptase inhibitor ( NRTI ), widely used to treat HIV infection. Zidovudine increases CRISPR/Cas9-mediated editing frequency. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2479 Supplier Medchemexpress LLC Supplier No. HY17413500MG
Medchemexpress, HY-17413 500mg Zidovudine Azidothymidine;AZT;ZDV CAS:30516-87-1 Formula:C10H13N5O4 NRTI, HIV CRISPR/Cas9 Purity:98% Zidovudine is a nucleoside reverse transcriptase inhibitor ( NRTI ), widely used to treat HIV infection. Zidovudine increases CRISPR/Cas9-mediated editing frequency. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.