Learn More
Medchemexpress LLC Zidovudine-d3 (azidothymidine-d3) | 30516-87-1 | 270.26 | 1 MG
SDP
Catalog No. 5000385299
Encompass_Preferred
Zidovudine-d3 is the deuterium labeled Zidovudine, a nucleoside reverse transcriptase inhibitor (NRTI) widely used to treat HIV infection.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules are used as tracers for quantitation during drug development.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-85299 Supplier Medchemexpress LLC Supplier No. HY17413S1MG
Zidovudine-d3 is the deuterium labeled Zidovudine, a nucleoside reverse transcriptase inhibitor (NRTI) widely used to treat HIV infection.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules are used as tracers for quantitation during drug development.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.