Learn More
Medchemexpress LLC Zidovudine-d4 | 30516-87-1 | 271.27 | 1 MG
SDP
Catalog No. 5000385967
Encompass_Preferred
Zidovudine-d4 is a deuterated labeled version of Zidovudine (HY-17413). Zidovudine is known as a nucleoside reverse transcriptase inhibitor (NRTI) and is extensively utilized in the treatment of HIV infection.
  • Can be employed as a tracer.
  • Serves as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-003-85967 Supplier Medchemexpress LLC Supplier No. HY17413S21MG
Zidovudine-d4 is a deuterated labeled version of Zidovudine (HY-17413). Zidovudine is known as a nucleoside reverse transcriptase inhibitor (NRTI) and is extensively utilized in the treatment of HIV infection.
  • Can be employed as a tracer.
  • Serves as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.