Learn More
Medchemexpress LLC Zidovudine-d4 (Azidothymidine-d4) | 30516-87-1 | 271.27 | C10H9D4N5O4 | 10 MG
SDP
Catalog No. 5000298701
Encompass_Preferred
Zidovudine-d4 (Azidothymidine-d4) is a deuterium-labeled analogue of zidovudine supplied for research and analytical applications. It is intended as an internal standard or tracer for mass spectrometry, quantitative assays, and method development in HIV-related studies. The labeled compound enables clear mass differentiation from the native drug during analysis.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for analytical quantitation and method development.
  • Provided as a 10 mg research-grade solid for laboratory use.
  • Molecular formula C10H9D4N5O4 and molecular weight 271.27 for accurate calculations.
  • Certificate of analysis available from the manufacturer for purity and quality control.
Catalog No. 50-002-98701 Supplier Medchemexpress LLC Supplier No. HY17413S210MG
Zidovudine-d4 (Azidothymidine-d4) is a deuterium-labeled analogue of zidovudine supplied for research and analytical applications. It is intended as an internal standard or tracer for mass spectrometry, quantitative assays, and method development in HIV-related studies. The labeled compound enables clear mass differentiation from the native drug during analysis.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for analytical quantitation and method development.
  • Provided as a 10 mg research-grade solid for laboratory use.
  • Molecular formula C10H9D4N5O4 and molecular weight 271.27 for accurate calculations.
  • Certificate of analysis available from the manufacturer for purity and quality control.