Learn More
Medchemexpress LLC Zifanocycline trifluoroacetate | 00-00-0 | 99.8% | 666.64 g/mol | C31H37F3N4O9 | 5 MG
SDP
Catalog No. 5000222219
Encompass_Preferred
Zifanocycline TFA is an orally active, semi-synthetic aminomethylcycline antibiotic supplied as the trifluoroacetate salt for research use. It inhibits bacterial ribosome function and demonstrates broad-spectrum in vitro activity against Gram-positive and Gram-negative bacteria, including multidrug-resistant strains. Supplied at high purity for laboratory studies and antimicrobial research applications.
  • Orally active aminomethylcycline antibiotic.
  • Broad-spectrum activity against gram-positive and gram-negative bacteria.
  • Useful for studying skin infections, community-acquired pneumonia, and intra-abdominal infections.
  • High purity, suitable for analytical and biological assays.
  • Provided as the trifluoroacetate (TFA) salt for improved handling.
  • Molecular weight 666.64 g/mol.
Catalog No. 50-002-22219 Supplier Medchemexpress LLC Supplier No. HY139554A5MG
Zifanocycline TFA is an orally active, semi-synthetic aminomethylcycline antibiotic supplied as the trifluoroacetate salt for research use. It inhibits bacterial ribosome function and demonstrates broad-spectrum in vitro activity against Gram-positive and Gram-negative bacteria, including multidrug-resistant strains. Supplied at high purity for laboratory studies and antimicrobial research applications.
  • Orally active aminomethylcycline antibiotic.
  • Broad-spectrum activity against gram-positive and gram-negative bacteria.
  • Useful for studying skin infections, community-acquired pneumonia, and intra-abdominal infections.
  • High purity, suitable for analytical and biological assays.
  • Provided as the trifluoroacetate (TFA) salt for improved handling.
  • Molecular weight 666.64 g/mol.