Learn More
Medchemexpress LLC HY-132001 5mg , Ziftomenib CAS:2134675-36-6 Purity:>98%
SDP
Catalog No. 502260718
Encompass_Preferred
HY-132001 5mg Ziftomenib CAS: 2134675-36-6 Ziftomenib (KO-539) is a menin-MLL interaction inhibitor with antitumor activities (WO2017161028A1, compound 151). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0718 Supplier Medchemexpress LLC Supplier No. HY1320015MG
HY-132001 5mg Ziftomenib CAS: 2134675-36-6 Ziftomenib (KO-539) is a menin-MLL interaction inhibitor with antitumor activities (WO2017161028A1, compound 151). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.