Learn More
Creative Biomart Recombinant Zika Envelope Domain-3 Antigen, His-tagged
Supplier:  Creative Biomart ZIK443
Encompass_Preferred
The E. coli derived Zika Envelope domain-III is a non-glycosylated polypeptide chain having a molecular mass of 11 kDa and fused to a His tag at N-terminus.
Catalog No. 50-208-4821