Learn More
Creative Biomart Recombinant Zika Envelope Antigen, His-tagged
Supplier:  Creative Biomart ZIK446
Encompass_Preferred
Sf9 derived recombinant Zika Envelope protein, having an Mw of approximately 50 kDa.The Zika Envelope protein is fused to a 6 His tag and purified by proprietary chromatographic technique.
Catalog No. 50-208-4824