Learn More
Creative Biomart Recombinant Zika Envelope Antigen, His-tagged
Supplier:  Creative Biomart ZIK442
Encompass_Preferred
The E. coli derived Recombinant Zika Envelope protein (250-410 a.a., Strain: ZikaSPH2015) having an Mw of 19 kDa is designed with minimal level cross-reaction to other Flavivirus such as dengue and We
Catalog No. 50-208-4820