Learn More
Enzo Life Sciences Zika Virus Envelope protein (Suriname strain) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2740100
Encompass_Preferred
Zika virus envelope protein recombinant antigen containing a C-terminal His-tag Zika virus envelope protein recombinant antigen containing a C-terminal His-tag Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 20mM NaH2PO4 pH 7.8, 210mM NaCl. Source: Produced in HEK293 mammalian cells. Recombinant Zika virus envelope protein with C-terminal 6-His tag. Long Term Storage: -80°C. Applications: ELISA Vaccine research, LINE assay, Antibody capture.
Catalog No. 50-201-4086