Learn More
Creative Biomart Recombinant Zika NS1 Antigen, His-tagged
Supplier:  Creative Biomart ZIK447
Encompass_Preferred
The E. coli derived Recombinant Zika NS1 protein (Strain: ZikaSPH2015) having an Mw of 45 kDa is derived from the full length Zika NS1 protein.The Zika NS1 protein is fused to a 6 His tag at C-term
Catalog No. 50-208-4825
 Restricted Item