Learn More
Creative Biomart Recombinant Zika NS1 Antigen, His-tagged
Supplier:  Creative Biomart ZIK448
Encompass_Preferred
The HEK derived recombinant Zika NS1 protein strain MR-766 (Prototype, Uganda, 1947, GenBank accession number AY632535).The Zika NS1 protein is fused to a 6 His tag at C-terminus and purified by pr
Catalog No. 50-208-4826