Learn More
Enzo Life Sciences Zika Virus -NS1 (Uganda strain) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2760100
Encompass_Preferred
Recombinant Zika Virus (Uganda strain) NS1 protein, incorporating a C-terminal 6x His tag produced in mammalian HEK293 cells. Recombinant Zika Virus (Uganda strain) NS1 protein, incorporating a C-terminal 6x His tag produced in mammalian HEK293 cells. Alternative name: ZIKV-NS1. Purity: ≥85% (SDS-PAGE). Formulation: Liquid. In DPBS pH 7.4. Source: Produced in mammalian HEK293 cells. Recombinant Zika Virus (Uganda strain) NS1 protein, incorporating a C-terminal 6x His tag. Long Term Storage: -80°C. Applications: ELISA Vaccine research, LINE assay, Antibody capture.
Catalog No. 50-201-4088