Learn More
Medchemexpress LLC Zika virus E (envelope) protein | 10 UG
SDP
Catalog No. 5000293819
Encompass_Preferred
Recombinant Zika virus envelope (E) protein fragment produced in HEK293 cells with a C-terminal His tag. Supplied as a 0.22 μm filtered PBS solution (pH 7.4), this reagent is intended for research applications such as antigenicity studies, ELISA, and antibody characterization. Store frozen at -80°C and avoid repeated freeze-thaw cycles.
  • Produced in HEK293 cells (human expression system).
  • C-terminal His tag for detection or affinity purification.
  • Supplied as a 0.22 μm filtered PBS solution, pH 7.4.
  • Approximate molecular weight 13 kDa.
  • Sequence corresponds to V593-K699 (accession ALU33341.1).
  • For research use only; not for diagnostic or therapeutic use.
  • Recommended long-term storage at -80°C; minimize freeze-thaw cycles.
Catalog No. 50-002-93819 Supplier Medchemexpress LLC Supplier No. HYP7445610UG
Recombinant Zika virus envelope (E) protein fragment produced in HEK293 cells with a C-terminal His tag. Supplied as a 0.22 μm filtered PBS solution (pH 7.4), this reagent is intended for research applications such as antigenicity studies, ELISA, and antibody characterization. Store frozen at -80°C and avoid repeated freeze-thaw cycles.
  • Produced in HEK293 cells (human expression system).
  • C-terminal His tag for detection or affinity purification.
  • Supplied as a 0.22 μm filtered PBS solution, pH 7.4.
  • Approximate molecular weight 13 kDa.
  • Sequence corresponds to V593-K699 (accession ALU33341.1).
  • For research use only; not for diagnostic or therapeutic use.
  • Recommended long-term storage at -80°C; minimize freeze-thaw cycles.