Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zika virus E/envelope protein (438 a.a., Sf9, His) | >90.0% | 10 UG
SDP

Catalog No. 5000292731
Encompass_Preferred

Recombinant Zika virus envelope (E) protein expressed in Sf9 insect cells as a partial sequence (residues F257-G694) with a C-terminal His tag. Supplied as a 0.22 μm filtered solution in Tris/NaCl/glycerol, the protein is >90% pure by reducing SDS-PAGE and intended for biochemical, immunological, and assay development applications. Store frozen and avoid repeated freeze-thaw cycles.

  • Partial E protein covering residues F257-G694.
  • Produced in Sf9 insect cells with a C-terminal His tag.
  • Supplied in Tris/NaCl/glycerol buffer, 0.22 μm filtered.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Predicted molecular mass 45.53 kDa.
  • Suitable for ELISA, antibody validation, and biochemical assays.
  • Store at -80°C and avoid repeated freeze-thaw cycles.

Catalog No. 50-002-92731 Supplier Medchemexpress LLC Supplier No. HYP7445810UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Zika virus envelope (E) protein expressed in Sf9 insect cells as a partial sequence (residues F257-G694) with a C-terminal His tag. Supplied as a 0.22 μm filtered solution in Tris/NaCl/glycerol, the protein is >90% pure by reducing SDS-PAGE and intended for biochemical, immunological, and assay development applications. Store frozen and avoid repeated freeze-thaw cycles.

  • Partial E protein covering residues F257-G694.
  • Produced in Sf9 insect cells with a C-terminal His tag.
  • Supplied in Tris/NaCl/glycerol buffer, 0.22 μm filtered.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Predicted molecular mass 45.53 kDa.
  • Suitable for ELISA, antibody validation, and biochemical assays.
  • Store at -80°C and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.