Learn More
Medchemexpress LLC Zika virus E/envelope protein | 5 UG
SDP
Catalog No. 5000293458
Encompass_Preferred
Recombinant Zika virus envelope (E) protein fragment (residues V593-K699) expressed in HEK293 cells with a His tag, supplied as a 0.22 μm filtered PBS solution for research use. The fragment is ~107 amino acids (~13 kDa) with endotoxin <1 EU/μg; store at -80°C and avoid repeated freeze-thaw cycles.
  • Expressed in HEK293 human cells for mammalian post-translational modifications.
  • His tag facilitates purification and detection.
  • Supplied as a sterile 0.22 μm filtered PBS solution.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Fragment corresponds to residues V593-K699 (≈107 aa, ~13 kDa).
  • Stable long-term at -80°C; avoid repeated freeze-thaw cycles.
Catalog No. 50-002-93458 Supplier Medchemexpress LLC Supplier No. HYP744565UG
Recombinant Zika virus envelope (E) protein fragment (residues V593-K699) expressed in HEK293 cells with a His tag, supplied as a 0.22 μm filtered PBS solution for research use. The fragment is ~107 amino acids (~13 kDa) with endotoxin <1 EU/μg; store at -80°C and avoid repeated freeze-thaw cycles.
  • Expressed in HEK293 human cells for mammalian post-translational modifications.
  • His tag facilitates purification and detection.
  • Supplied as a sterile 0.22 μm filtered PBS solution.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Fragment corresponds to residues V593-K699 (≈107 aa, ~13 kDa).
  • Stable long-term at -80°C; avoid repeated freeze-thaw cycles.