Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zika virus E/envelope protein (HEK293, His) | >95.0% | 5 UG
SDP

Catalog No. 5000292151
Encompass_Preferred

Recombinant Zika virus E (envelope) protein expressed in HEK293 cells, supplied lyophilized for use in immunoassays, antibody validation, and serology research. The protein includes a C-terminal His tag and corresponds to envelope domain V593-K699 (≈13 kDa), provided in small-format quantities suitable for assay development.

  • Recombinant expression in HEK293 mammalian cells.
  • C-terminal His tag for detection and purification.
  • Domain V593-K699 of the envelope protein, sequence verified.
  • Purity >95% by reducing SDS-PAGE.
  • Lyophilized formulation from PBS, pH 7.4.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week.

Catalog No. 50-002-92151 Supplier Medchemexpress LLC Supplier No. HYP761295UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Zika virus E (envelope) protein expressed in HEK293 cells, supplied lyophilized for use in immunoassays, antibody validation, and serology research. The protein includes a C-terminal His tag and corresponds to envelope domain V593-K699 (≈13 kDa), provided in small-format quantities suitable for assay development.

  • Recombinant expression in HEK293 mammalian cells.
  • C-terminal His tag for detection and purification.
  • Domain V593-K699 of the envelope protein, sequence verified.
  • Purity >95% by reducing SDS-PAGE.
  • Lyophilized formulation from PBS, pH 7.4.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.