Learn More
Medchemexpress LLC Zika virus E (envelope) protein (residues F257-G694) | >90.0% | 50 UG
SDP
Catalog No. 5000293817
Encompass_Preferred
Recombinant Zika virus E (envelope) protein (residues F257-G694) expressed in Sf9 insect cells with a C-terminal His tag. Supplied as a 0.22 μm filtered solution formulated in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0; intended for research use, shipped on dry ice, and stored at -80°C.
  • Protein corresponds to residues F257-G694 of the envelope protein.
  • Expressed in Sf9 insect cells for eukaryotic folding and post-translational modifications.
  • C-terminal His tag enables affinity purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Formulated in Tris/NaCl/glycerol buffer; exchange buffer for SPR assays is recommended.
  • Shipped on dry ice; store at -80°C and avoid repeated freeze-thaw cycles.
Catalog No. 50-002-93817 Supplier Medchemexpress LLC Supplier No. HYP7445850UG
Recombinant Zika virus E (envelope) protein (residues F257-G694) expressed in Sf9 insect cells with a C-terminal His tag. Supplied as a 0.22 μm filtered solution formulated in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0; intended for research use, shipped on dry ice, and stored at -80°C.
  • Protein corresponds to residues F257-G694 of the envelope protein.
  • Expressed in Sf9 insect cells for eukaryotic folding and post-translational modifications.
  • C-terminal His tag enables affinity purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Formulated in Tris/NaCl/glycerol buffer; exchange buffer for SPR assays is recommended.
  • Shipped on dry ice; store at -80°C and avoid repeated freeze-thaw cycles.