Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zika virus envelope (E) protein | >90.0% | 100 UG
SDP

Catalog No. 5000293454
Encompass_Preferred

Recombinant Zika virus envelope (E) protein, full-length (438 amino acids), produced in Sf9 insect cells with a C-terminal His tag. Supplied as a 0.22 μm filtered buffered solution (20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0) for research use only. Purity is reported as greater than 90% by reducing SDS-PAGE. Store at -80°C, aliquot to avoid repeated freeze-thaw cycles.

  • Recombinant full-length envelope protein (438 a.a.).
  • Expressed in Sf9 insect cells for eukaryotic processing.
  • C-terminal His tag facilitates purification and detection.
  • Supplied in a ready-to-use buffered formulation.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Suitable for immunoassays, antibody production, and biochemical characterization.
  • Store at -80°C; avoid repeated freeze-thaw cycles.

Catalog No. 50-002-93454 Supplier Medchemexpress LLC Supplier No. HYP74458100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Zika virus envelope (E) protein, full-length (438 amino acids), produced in Sf9 insect cells with a C-terminal His tag. Supplied as a 0.22 μm filtered buffered solution (20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0) for research use only. Purity is reported as greater than 90% by reducing SDS-PAGE. Store at -80°C, aliquot to avoid repeated freeze-thaw cycles.

  • Recombinant full-length envelope protein (438 a.a.).
  • Expressed in Sf9 insect cells for eukaryotic processing.
  • C-terminal His tag facilitates purification and detection.
  • Supplied in a ready-to-use buffered formulation.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Suitable for immunoassays, antibody production, and biochemical characterization.
  • Store at -80°C; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.