Learn More
Cayman Chemical ZIka VIrus Envelope ProteIn
SDP
Catalog No. 502944876
Encompass_Preferred
Source: Recombinant Zika virus C-terminal His-tagged E protein expressed in insect cells • Amino acids: 291-696 • MW: 45.6 kDa
Catalog No. 50-294-4876 Supplier Cayman Chemical Supplier No. 41068100 UG
Source: Recombinant Zika virus C-terminal His-tagged E protein expressed in insect cells • Amino acids: 291-696 • MW: 45.6 kDa