Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Zika Virus Ns1 (Q32Ze1, 792 A.A.-1144 A.A.) Partial Recombinant Protein Expressed In Baculovirus Expression System

Catalog No. 89033710 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-710 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-710 Supplier Abnova™ Supplier No. P6205
Only null left
Add to Cart
Add to Cart
High quality

Specifications

Accession Number Q32ZE1
For Use With (Application) ELISA, Lateral-Flow Immunochromatographic Assay, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 7751225
Name NS1 (Zika virus) Recombinant Protein
Preparation Method Baculovirus expression system
Quality Control Testing Coomassie stained SDS-PAGE gel
Quantity 100 μg
Storage Requirements Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Species Baculovirus
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.