Learn More
Cayman Chemical ZIka VIrus NS5 ProteInstraI
SDP
Catalog No. 502944726
Encompass_Preferred
Source: Recombinant Zika virus C-Terminal His-tagged NS5 (strain Zika SPH2015) expressed in insect cells • Amino acids: 2,772-3,423 • MW: 76.7 kDa
Catalog No. 50-294-4726 Supplier Cayman Chemical Supplier No. 41079100 UG
Source: Recombinant Zika virus C-Terminal His-tagged NS5 (strain Zika SPH2015) expressed in insect cells • Amino acids: 2,772-3,423 • MW: 76.7 kDa