Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Zika Virus (Strain: MR 766) Infectious Culture Fluid
SDP

Catalog No. 22158080 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-080 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order
Catalog No. 22-158-080 Supplier ZeptoMetrix Supplier No. 0810521CF
Add to Cart
Add to Cart
Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order

Used for research and product development testing. ZeptoMetrix Zika virus (Strain: MR 766) Infectious Culture Fluid is sold as consumable testing materials.

  • Enveloped, icosahedral Flavivirus that contains a non-segmented, single-stranded positive-sense RNA
  • Propagated in the Vero or LLC-mk2 cell line
  • Assayed for its Tissue Culture Infective Dose(TCID50), and sold with titers >105U/mL

Specifications

Biosafety Level 2
Certifications/Compliance ISO 13485
Content And Storage -65°C or below
Form Solution
Media Type Vial
Product Type Zika Virus Infectious Culture Fluid
For Use With (Application) Nucleic acid/molecular testing, limit of detection (LOD) studies, cross-reactivity studies, other viral-based assays
Quantity 1 mL

For research use only. Not for use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.