Learn More
ATCC Zika virus; Strain: PRVABC59
Catalog No. 502381981
Encompass
Zika virus strain PRVABC59 is propagated in VERO C1008 cells [Vero 76, clone E6, Vero E6] cells (ATCC CRL-1586). This strain was isolated in 2015 from a human serum specimen in Puerto Rico and was deposited by the Centers for Disease Control and Prevention. This viral strain has applications in infectious disease, vector-borne disease, and zoonotic disease research.
Catalog No. 50-238-1981 Supplier ATCC Supplier No. VR1843
Zika virus strain PRVABC59 is propagated in VERO C1008 cells [Vero 76, clone E6, Vero E6] cells (ATCC CRL-1586). This strain was isolated in 2015 from a human serum specimen in Puerto Rico and was deposited by the Centers for Disease Control and Prevention. This viral strain has applications in infectious disease, vector-borne disease, and zoonotic disease research.