Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Zika Virus (Strain: PRVABC59) Lysate
SDP

Catalog No. 22156116 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-116 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-116 Supplier ZeptoMetrix Supplier No. 0810525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be used as an antigen, as a source for the purification of viral proteins, or for the detection of viral antibodies

Zika virus is an enveloped, icosahedral Flavivirus that contains a non-segmented, single-stranded positive-sense RNA.

  • Propagated in the Vero of LLC-mk2 cell line
  • Purified using sucrose density gradient ultracentrifugation, disrupted in the presence of 0.5% Triton X-100 non-ionic detergent/0.6M KCl, and heat inactivated

Specifications

Biosafety Level 2
Content And Storage -65°C or below
For Use With (Application) EIA, Western blot, Dot blot, Other protein-based assays
Quantity 1 mg

Do not use in humans. For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.