Learn More
Enzo Life Sciences Zika Virus VLP (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2780100
Encompass_Preferred
Recombinant Zika virus-like particles consisting of Envelope, pre-Membrane, and Membrane protein produced in mammalian HEK293 cells. Recombinant Zika virus-like particles consisting of Envelope, pre-Membrane, and Membrane protein produced in mammalian HEK293 cells. Alternative name: ZIKV-VLP. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In DPBS pH 7.4, 20% Sucrose. May contain traces of tartrate. Source: Produced in mammalian HEK293 cells. Recombinant Zika virus-like particles consisting of envelope, pre-membrane, and membrane protein. Long Term Storage: -80°C. Applications: ELISA Vaccine research, LINE assay, Antibody capture.
Catalog No. 50-201-4090