Learn More
Sino Biological ZIKV ZIKV-E MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40543MM3520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Zika virus (strain Zika SPH2015) Envelope Protein (ZIKV-E) ( Catalog#40543-V08H; ALU33341.1; Val593-Lys699). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2849